2,4-Dimethylquinoline, 95%, Thermo Scientific Chemicals
2,4-Dimethylquinoline, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4-Dimethylquinoline, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
100 mg
Catalog number L08348.ME
also known as L08348-ME
Price (USD)/ Each
80.70
-
Add to cart
Quantity:
500 mg
Request bulk or custom format
Price (USD)/ Each
80.70
Add to cart
2,4-Dimethylquinoline, 95%, Thermo Scientific Chemicals
Catalog numberL08348.ME
Price (USD)/ Each
80.70
-
Add to cart
Chemical Identifiers
CAS1198-37-4
IUPAC Name2,4-dimethylquinoline
Molecular FormulaC11H11N
InChI KeyZTNANFDSJRRZRJ-UHFFFAOYSA-N
SMILESCC1=CC(C)=C2C=CC=CC2=N1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to yellow
Assay (GC)≥94.0%
Refractive Index1.6040-1.6100 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only