Thermo Scientific Chemicals

Triethyl methanetricarboxylate, 98%, Thermo Scientific Chemicals

Catalog number: L08356.06
5 g, Each
Thermo Scientific Chemicals

Triethyl methanetricarboxylate, 98%, Thermo Scientific Chemicals

Catalog number: L08356.06
5 g, Each
Quantity
Catalog number: L08356.06
also known as L08356-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6279-86-3
IUPAC Name
triethyl methanetricarboxylate
Molecular Formula
C10H16O6
InChI Key
AGZPNUZBDCYTBB-UHFFFAOYSA-N
SMILES
CCOC(=O)C(C(=O)OCC)C(=O)OCC
Appearance (Color)
Colorless to white to pale yellow
Form
Crystals or powder or crystalline powder or fused solid or clear liquid or viscous liquid as melt
Identification (FTIR)
Conforms
Refractive Index
1.4235-1.4275 @ 20?C
Assay (GC)
≥97.5%

Description

Methanetricarboxylates have been used as blocked malonic esters, restricting reaction with alkyl halides to monoalkylation. The unwanted carboxyl group can be removed by treatment with Na alkoxide, LDA or BCl3. Provides a useful 2-carbon chain extension method that adds to Michael acceptors under phase-transfer conditions. Triethyl Methanetricarboxylate is used in the synthesis of novel inhibitors of Hsp90. It is also used in the preparation of novel dihydroquinoline-3-carboxylic acids that function as HIV-1 integrase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methanetricarboxylates have been used as “blocked” malonic esters, restricting reaction with alkyl halides to monoalkylation. The unwanted carboxyl group can be removed by treatment with Na alkoxide, LDA or BCl3. Provides a useful 2-carbon chain extension method that adds to Michael acceptors under phase-transfer conditions. Triethyl Methanetricarboxylate is used in the synthesis of novel inhibitors of Hsp90. It is also used in the preparation of novel dihydroquinoline-3-carboxylic acids that function as HIV-1 integrase inhibitors.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text