Thermo Scientific Chemicals

1-Cyanoacetyl-3,5-dimethyl-1H-pyrazole, 97%, Thermo Scientific Chemicals

Catalog number: L08369.06
5 g, Each
Thermo Scientific Chemicals

1-Cyanoacetyl-3,5-dimethyl-1H-pyrazole, 97%, Thermo Scientific Chemicals

Catalog number: L08369.06
5 g, Each
Quantity
Catalog number: L08369.06
also known as L08369-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
36140-83-7
IUPAC Name
3-(3,5-dimethyl-1H-pyrazol-1-yl)-3-oxopropanenitrile
Molecular Formula
C8H9N3O
InChI Key
DDWZYWSLHBDVGR-UHFFFAOYSA-N
SMILES
CC1=NN(C(=O)CC#N)C(C)=C1
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
≤2.5%
Proton NMR
Conforms to structure
Appearance (Color)
White to cream
Form
Crystalline powder

Description

1-Cyanoacetyl-3,5-dimethyl-1H-pyrazole is used in pharma industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Cyanoacetyl-3,5-dimethyl-1H-pyrazole is used in pharma industry.

Notes
Hygroscopic. Store away from water and oxidizing agents. Protect from humidity and water. Incompatible with oxidizing agents, Strong acids, Strong bases, Strong reducing agents, Acid chlorides, Acid anhydrides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text