Thermo Scientific Chemicals

2,3-Dimethylnaphthalene, 97%, Thermo Scientific Chemicals

Catalog number: L08386.06
5 g, Each
Thermo Scientific Chemicals

2,3-Dimethylnaphthalene, 97%, Thermo Scientific Chemicals

Catalog number: L08386.06
5 g, Each
Quantity
Catalog number: L08386.06
also known as L08386-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
581-40-8
IUPAC Name
2,3-dimethylnaphthalene
Molecular Formula
C12H12
InChI Key
WWGUMAYGTYQSGA-UHFFFAOYSA-N
SMILES
CC1=C(C)C=C2C=CC=CC2=C1
Appearance (Color)
White to pale cream to black
Form
Crystals or powder or crystalline powder or flakes
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
100.0-109.0?C

Description

It is used in the identification of individual aromatic hydrocarbons in kerosene fraction (b.p. 150-250°).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the identification of individual aromatic hydrocarbons in kerosene fraction (b.p. 150-250°).

Solubility
Solubility in toluene (almost transparency). Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text