cis-6-Nonen-1-ol, 95%, Thermo Scientific Chemicals
cis-6-Nonen-1-ol, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

cis-6-Nonen-1-ol, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L08387.03
also known as L08387-03
Price (USD)/ Each
35.50
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
35.50
Add to cart
cis-6-Nonen-1-ol, 95%, Thermo Scientific Chemicals
Catalog numberL08387.03
Price (USD)/ Each
35.50
-
Add to cart
Chemical Identifiers
CAS35854-86-5
IUPAC Name(6Z)-non-6-en-1-ol
Molecular FormulaC9H18O
InChI KeyXJHRZBIBSSVCEL-ARJAWSKDSA-N
SMILESCCC=C/CCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥94.0%
Refractive Index1.4480-1.4540 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only