Thermo Scientific Chemicals

2-Amino-1,3-propanediol, 98%, Thermo Scientific Chemicals

Catalog number: L08426.06
5 g, Each
Thermo Scientific Chemicals

2-Amino-1,3-propanediol, 98%, Thermo Scientific Chemicals

Catalog number: L08426.06
5 g, Each
Quantity
Catalog number: L08426.06
also known as L08426-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
534-03-2
IUPAC Name
2-aminopropane-1,3-diol
Molecular Formula
C3H9NO2
InChI Key
KJJPLEZQSCZCKE-UHFFFAOYSA-N
SMILES
NC(CO)CO
Water Content (Karl Fischer Titration)
<2%
Appearance (Color)
White
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%
Form
Crystals or Crystalline powder
Melting Point
51-57?C

Description

2-Amino-1,3-propanediol is used as an intermediate in several chemical applications and also useful for serinol and some of its derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-1,3-propanediol is used as an intermediate in several chemical applications and also useful for serinol and some of its derivatives.

Solubility
Soluble in water and alcohol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, strong acids and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text