6-Benzoylhexanoic acid, 94%, Thermo Scientific Chemicals
6-Benzoylhexanoic acid, 94%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Benzoylhexanoic acid, 94%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Catalog number L08431.03
also known as L08431-03
Price (USD)/ Each
59.65
Online exclusive
66.00 
Save 6.35 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
59.65
Online exclusive
66.00 
Save 6.35 (10%)
Add to cart
6-Benzoylhexanoic acid, 94%, Thermo Scientific Chemicals
Catalog numberL08431.03
Price (USD)/ Each
59.65
Online exclusive
66.00 
Save 6.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7472-43-7
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point (clear melt)79.0-88.0?C
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥92.0 to ≤108.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only