Thermo Scientific Chemicals

Norbornane-2-carboxylic acid, 98%, predominantly endo isomer, Thermo Scientific Chemicals

Catalog number: L08439.06
5 g, Each
Thermo Scientific Chemicals

Norbornane-2-carboxylic acid, 98%, predominantly endo isomer, Thermo Scientific Chemicals

Catalog number: L08439.06
5 g, Each
Quantity
Catalog number: L08439.06
also known as L08439-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
824-62-4
IUPAC Name
bicyclo[2.2.1]heptane-2-carboxylic acid
Molecular Formula
C8H12O2
InChI Key
JESWDXIHOJGWBP-UHFFFAOYNA-N
SMILES
OC(=O)C1CC2CCC1C2
Assay (GC)
≥97.5% (sum of endo and exo isomers)
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Form
Crystals or waxy solid
Identification (FTIR)
Conforms
Appearance (Color)
White

Description

Norbornane-2-carboxylic acid is used in the preparation of 2-Bromonorbornane-1-carboxylic acid. It is also used as Pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Norbornane-2-carboxylic acid is used in the preparation of 2-Bromonorbornane-1-carboxylic acid. It is also used as Pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents and strong bases. Incompatible with oxidizing agents, Strong acids, Strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text