5-Norbornene-2-carboxylic acid is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Norbornene-2-carboxylic acid is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
Solubility
Not miscible with water.
Notes
Incompatible with oxidizing agents, bases. Store away from oxidizing agents.
RUO – Research Use Only
Benjamin D Fairbanks; Michael P Schwartz; Alexandra E Halevi; Charles R Nuttelman; Christopher N Bowman; Kristi S Anseth. A Versatile Synthetic Extracellular Matrix Mimic via Thiol-Norbornene Photopolymerization.Advanced Materials. 2009, 21 (48), 5005-5010.