2,4,6-Triisopropylbenzenesulfonamide, 98%, Thermo Scientific Chemicals
2,4,6-Triisopropylbenzenesulfonamide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4,6-Triisopropylbenzenesulfonamide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L08445.14
also known as L08445-14
Price (USD)/ Each
217.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
217.00
Add to cart
2,4,6-Triisopropylbenzenesulfonamide, 98%, Thermo Scientific Chemicals
Catalog numberL08445.14
Price (USD)/ Each
217.00
-
Add to cart
Chemical Identifiers
CAS105536-22-9
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)115.0-121.0?C
Appearance (Color)White
Assay (HPLC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only