Thermo Scientific Chemicals

N-Boc-L-aspartic acid, 98%, Thermo Scientific Chemicals

Catalog number: L08498.03
1 g, Each
Thermo Scientific Chemicals

N-Boc-L-aspartic acid, 98%, Thermo Scientific Chemicals

Catalog number: L08498.03
1 g, Each
Quantity
Catalog number: L08498.03
also known as L08498-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13726-67-5
IUPAC Name
2-{[(tert-butoxy)carbonyl]amino}butanedioic acid
Molecular Formula
C9H15NO6
InChI Key
KAJBMCZQVSQJDE-UHFFFAOYNA-N
SMILES
CC(C)(C)OC(=O)NC(CC(O)=O)C(O)=O
Appearance (Color)
White to pale cream
Water Content (Karl Fischer Titration)
≤2% (non-U.S. sourced material)
Form
Powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5% (non-U.S. sourced material)
Comment
Material Sourced in the U.S. and in other countries

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text