Thermo Scientific Chemicals

tert-Butyl hydrogen phthalate, 97%, Thermo Scientific Chemicals

Catalog number: L08534.14
25 g, Each
Thermo Scientific Chemicals

tert-Butyl hydrogen phthalate, 97%, Thermo Scientific Chemicals

Catalog number: L08534.14
25 g, Each
Quantity
Catalog number: L08534.14
also known as L08534-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
33693-84-4
Form
Crystals or powder or crystalline powder or granules
Appearance (Color)
White to cream to yellow
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Melting Point (clear melt)
81.0-91.0?C
Assay (Silylated GC)
≥96.0%

Description

It finds its application in the resolution and reactions of tertiary alcohols : 2-phenylbutan-2-ol and 3-methylhexan-3-ol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the resolution and reactions of tertiary alcohols : 2-phenylbutan-2-ol and 3-methylhexan-3-ol.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text