N2-Boc-D-glutamine is an N-Boc-protected form of D-Glutamine. D-Glutamine is an unnatural isomer of L-Glutamine that is present in human plasma an is a source of liberated ammonia. D-Glutamine can be synthesized by enzymatic means or can be found in cheeses, wine and vinegars as well.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N2-Boc-D-glutamine is an N-Boc-protected form of D-Glutamine. D-Glutamine is an unnatural isomer of L-Glutamine that is present in human plasma an is a source of liberated ammonia. D-Glutamine can be synthesized by enzymatic means or can be found in cheeses, wine and vinegars as well.
Solubility
Soluble, 1mmol in 2mL DMF. Soluble in chloroform and methanol.
Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
RUO – Research Use Only
Philip S. Portoghese, et al. Hybrid bivalent ligands with opiate and enkephalin pharmacophores.J. Med. Chem.,1987,30(11), 1991-1994.
A. N. Chulin, et al. Preparation and reactivity of aminoacyl pyroglutamates. Facile synthesis of 10-membered-ring cyclic dipeptides derived from 1,4-diaminobutyric and glutamic acids.Journal of Peptide Science.,2005,11(3), 175-186.