Thermo Scientific Chemicals

(R)-(-)-S-Methyl-S-phenylsulfoximine, 97%, Thermo Scientific Chemicals

Catalog number: L08562.03
1 g, Each
Thermo Scientific Chemicals

(R)-(-)-S-Methyl-S-phenylsulfoximine, 97%, Thermo Scientific Chemicals

Catalog number: L08562.03
1 g, Each
Quantity
Catalog number: L08562.03
also known as L08562-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
60933-65-5
IUPAC Name
imino(methyl)phenyl-λ⁶-sulfanone
Molecular Formula
C7H9NOS
InChI Key
YFYIDTVGWCYSEO-UHFFFAOYNA-N
SMILES
CS(=N)(=O)C1=CC=CC=C1
Form
Crystals or powder or crystalline powder or lumps or fused solid or clear liquid or viscous liquid as melt
Assay (GC)
≥96.0%
Appearance (Color)
White to pale yellow
Melting Point (clear melt)
25.0-35.0?C
Optical Rotation
-34? to -39.5? (c=1, acetone)

Description

(R)-(-)-S-Methyl-S-phenylsulfoximine is used as chiral ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-S-Methyl-S-phenylsulfoximine is used as chiral ligands.

Notes
Hygroscopic. Store away from water and oxidizing agents. Protect from humidity and water. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text