(R)-(-)-S-Methyl-S-phenylsulfoximine, 97%, Thermo Scientific Chemicals
(R)-(-)-S-Methyl-S-phenylsulfoximine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(R)-(-)-S-Methyl-S-phenylsulfoximine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L08562.MD
also known as L08562-MD
Price (USD)/ Each
96.80
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
96.80
Add to cart
(R)-(-)-S-Methyl-S-phenylsulfoximine, 97%, Thermo Scientific Chemicals
Catalog numberL08562.MD
Price (USD)/ Each
96.80
-
Add to cart
Chemical Identifiers
CAS60933-65-5
IUPAC Nameimino(methyl)phenyl-λ⁶-sulfanone
Molecular FormulaC7H9NOS
InChI KeyYFYIDTVGWCYSEO-UHFFFAOYNA-N
SMILESCS(=N)(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or lumps or fused solid or clear liquid or viscous liquid as melt
Assay (GC)≥96.0%
Appearance (Color)White to pale yellow
Melting Point (clear melt)25.0-35.0?C
Optical Rotation-34? to -39.5? (c=1, acetone)
View more
(R)-(-)-S-Methyl-S-phenylsulfoximine is used as chiral ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-S-Methyl-S-phenylsulfoximine is used as chiral ligands.

Notes
Hygroscopic. Store away from water and oxidizing agents. Protect from humidity and water. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Carsten Bolm; Marinella Verrucci; Oliver Simic; Pier Giorgio Cozzi; Gerhard Raabe; Hiroaki Okamura. A new class of C1-symmetric monosulfoximine ligands for enantioselective hetero Diels-Alder reactions.Chemical Communications.2003, (22), 2826-2827.
  2. Stephen G. Pyne; Branko Dikic. Diastereoselective additions of lithiated N-tertbutyldiphenylsilyl- S-methyl-S-phenylsulfoximine to imines and aldehydes.Tetrahedron Letters.1990, 31 5231-5234.