Thermo Scientific Chemicals

3-Chloro-N,N-dimethylaniline, 95%, Thermo Scientific Chemicals

Catalog number: L08563.06
5 g, Each
Thermo Scientific Chemicals

3-Chloro-N,N-dimethylaniline, 95%, Thermo Scientific Chemicals

Catalog number: L08563.06
5 g, Each
Quantity
Catalog number: L08563.06
also known as L08563-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6848-13-1
IUPAC Name
3-chloro-N,N-dimethylaniline
Molecular Formula
C8H10ClN
InChI Key
CHHCCYVOJBBCIY-UHFFFAOYSA-N
SMILES
CN(C)C1=CC=CC(Cl)=C1
Refractive Index
1.5700-1.5780 @ 20?C
Form
Liquid
Appearance (Color)
Clear colorless to yellow to orange or pale brown
Identification (FTIR)
Conforms
Assay (GC)
≥94.0%

Description

It can be used to produce 3-(3-dimethylamino-phenyl)-acrylic acid butyl ester at the temperature of 140°C in the presence of reagent Na2CO3 and solvent N,N-dimethyl-acetamide with the reaction time of 20 hours.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used to produce 3-(3-dimethylamino-phenyl)-acrylic acid butyl ester at the temperature of 140°C in the presence of reagent Na2CO3 and solvent N,N-dimethyl-acetamide with the reaction time of 20 hours.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text