Thermo Scientific Chemicals

Trimethyl-1-octylammonium bromide, 97%, Thermo Scientific Chemicals

Catalog number: L08610.14
25 g, Each
Thermo Scientific Chemicals

Trimethyl-1-octylammonium bromide, 97%, Thermo Scientific Chemicals

Catalog number: L08610.14
25 g, Each
Quantity
Catalog number: L08610.14
also known as L08610-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2083-68-3
IUPAC Name
trimethyl(octyl)azanium bromide
Molecular Formula
C11H26BrN
InChI Key
XCOHAFVJQZPUKF-UHFFFAOYSA-M
SMILES
[Br-].CCCCCCCC[N+](C)(C)C
Assay (Titration ex Bromide)
≥96.0 to ≤104.0%
Water Content (Karl Fischer Titration)
≤2.25%
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder or lumps or fused solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text