4-Phenylcyclohexanone, 98+%, Thermo Scientific Chemicals
4-Phenylcyclohexanone, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Phenylcyclohexanone, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number L08614.22
also known as L08614-22
Price (USD)/ Each
309.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
309.00
Add to cart
4-Phenylcyclohexanone, 98+%, Thermo Scientific Chemicals
Catalog numberL08614.22
Price (USD)/ Each
309.00
-
Add to cart
Chemical Identifiers
CAS4894-75-1
IUPAC Name4-phenylcyclohexan-1-one
Molecular FormulaC12H14O
InChI KeyYKAYMASDSHFOGI-UHFFFAOYSA-N
SMILESO=C1CCC(CC1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (GC)≥98.0%
Melting Point (clear melt)76.0-82.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only