4-n-Decylaniline, 98%, Thermo Scientific Chemicals
4-n-Decylaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-n-Decylaniline, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L08620.03
also known as L08620-03
Price (USD)/ Each
38.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
38.00
Add to cart
4-n-Decylaniline, 98%, Thermo Scientific Chemicals
Catalog numberL08620.03
Price (USD)/ Each
38.00
-
Add to cart
Chemical Identifiers
CAS37529-30-9
IUPAC Name4-decylaniline
Molecular FormulaC16H27N
InChI KeyWGENWPANMZLPIH-UHFFFAOYSA-N
SMILESCCCCCCCCCCC1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to yellow to orange to red to brown
FormCrystals or powder or crystalline powder or lumps or chunks or fused solid or clear liquid as melt
Assay (GC)≥97.5%
Refractive Index1.5085-1.5125 @ 20?C
It is used to synthesize p,p'-Di-n-decyl-azobenzol could be obtained by the reactant of 4-Decylaniline, and the reagents of O2, CuCl, pyridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to synthesize p,p′-Di-n-decyl-azobenzol could be obtained by the reactant of 4-Decylaniline, and the reagents of O2, CuCl, pyridine.

Solubility
Insoluble in water.

Notes
Air sensitive. Store away air and from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.
RUO – Research Use Only

General References:

  1. Channabasaveshwar V. Yelamaggad, et al. The biaxial smectic (SmAb) phase in nonsymmetric liquid crystal dimers comprising two rodlike anisometric segments: an unusual behavior.J. Mater. Chem.,2006,16(42), 4099-4102.
  2. Channabasaveshwar V. Yelamaggad, et al. Self-Assembly of C3h and Cs Symmetric Keto-enamine Forms of Tris(N-salicyl-ideneanilines) into Columnar Phases: A New Family of Discotic Liquid Crystals.J. Am. Chem. Soc.,2004,126(21), 6506-6507.