Thermo Scientific Chemicals

4-n-Decylaniline, 98%, Thermo Scientific Chemicals

Catalog number: L08620.14
25 g, Each
Thermo Scientific Chemicals

4-n-Decylaniline, 98%, Thermo Scientific Chemicals

Catalog number: L08620.14
25 g, Each
Quantity
Catalog number: L08620.14
also known as L08620-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
37529-30-9
IUPAC Name
4-decylaniline
Molecular Formula
C16H27N
InChI Key
WGENWPANMZLPIH-UHFFFAOYSA-N
SMILES
CCCCCCCCCCC1=CC=C(N)C=C1
Appearance (Color)
Colorless to white to yellow to orange to red to brown
Form
Crystals or powder or crystalline powder or lumps or chunks or fused solid or clear liquid as melt
Assay (GC)
≥97.5%
Refractive Index
1.5085-1.5125 @ 20?C

Description

It is used to synthesize p,p'-Di-n-decyl-azobenzol could be obtained by the reactant of 4-Decylaniline, and the reagents of O2, CuCl, pyridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to synthesize p,p′-Di-n-decyl-azobenzol could be obtained by the reactant of 4-Decylaniline, and the reagents of O2, CuCl, pyridine.

Solubility
Insoluble in water.

Notes
Air sensitive. Store away air and from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text