1,2-Dibenzoylethane is effective in inhibiting the mammary DMBA-DNA adduct formation. This inhibitory effect on mammary DNA adduct formation was associated with increased liver activities of glutathione S-transferase, QR, and 7-ethoxyresorufin-O-deethylase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,2-Dibenzoylethane is effective in inhibiting the mammary DMBA-DNA adduct formation. This inhibitory effect on mammary DNA adduct formation was associated with increased liver activities of glutathione S-transferase, QR, and 7-ethoxyresorufin-O-deethylase.
Solubility
Soluble in acetone. Insoluble in water.
Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only
Gary W. Griffin; Robert B. Hager. Pinacol Reduction of 1,2-Dibenzoylethane. Formation of a Strained Cyclic Pinacol.J. Org. Chem.1963, 28 (2), 599-600.