Thermo Scientific Chemicals

L-Norvaline, 99%

Catalog number: L08658.06
5 g, Each
Thermo Scientific Chemicals

L-Norvaline, 99%

Catalog number: L08658.06
5 g, Each
Quantity
Catalog number: L08658.06
also known as L08658-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6600-40-4
IUPAC Name
(2S)-2-aminopentanoic acid
Molecular Formula
C5H11NO2
InChI Key
SNDPXSYFESPGGJ-BYPYZUCNSA-N
SMILES
CCC[C@H](N)C(O)=O
Optical Rotation
24.0 ± 1.5? (C=10 in 5N HCl)
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%
Appearance (Color)
White
Form
Crystals or powder or crystalline powder or fused solid

Description

L-Norvaline enhances NO production from activated macrophages. It is known to promote tissue regeneration and muscle growth and become a precursor in the penicillin biosynthetic pathway. It also can be used for nutrition agent and drug synthesis. For example: it can react with phthalic acid anhydride to get N,N-phthaloyl-L-norvaline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Norvaline enhances NO production from activated macrophages. It is known to promote tissue regeneration and muscle growth and become a precursor in the penicillin biosynthetic pathway. It also can be used for nutrition agent and drug synthesis. For example: it can react with phthalic acid anhydride to get N,N-phthaloyl-L-norvaline.

Solubility
Soluble in water (almost transparency), 10.5 g/100 mL (18°C).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text