Erucic acid, tech. 90%, Thermo Scientific Chemicals
Erucic acid, tech. 90%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Erucic acid, tech. 90%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number L08685.30
also known as L08685-30
Price (USD)/ Each
117.65
Online exclusive
130.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
117.65
Online exclusive
130.00 
Save 12.35 (10%)
Add to cart
Erucic acid, tech. 90%, Thermo Scientific Chemicals
Catalog numberL08685.30
Price (USD)/ Each
117.65
Online exclusive
130.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS112-86-7
IUPAC Name(13Z)-docos-13-enoic acid
Molecular FormulaC22H42O2
InChI KeyDPUOLQHDNGRHBS-KTKRTIGZSA-N
SMILESCCCCCCCCC=C/CCCCCCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
Formfused solid, clear as melt
Assay (Silylated GC)> 88.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only