Ethyl indole-3-acrylate, 97%, Thermo Scientific Chemicals
Ethyl indole-3-acrylate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl indole-3-acrylate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L08703.06
also known as L08703-06
Price (USD)/ Each
141.65
Online exclusive
157.00 
Save 15.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
141.65
Online exclusive
157.00 
Save 15.35 (10%)
Add to cart
Ethyl indole-3-acrylate, 97%, Thermo Scientific Chemicals
Catalog numberL08703.06
Price (USD)/ Each
141.65
Online exclusive
157.00 
Save 15.35 (10%)
-
Add to cart
Chemical Identifiers
CAS15181-86-9
IUPAC Nameethyl 3-(1H-indol-3-yl)prop-2-enoate
Molecular FormulaC13H13NO2
InChI KeyOQJSITNIWIYWPU-UHFFFAOYSA-N
SMILESCCOC(=O)C=CC1=CNC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥96.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)116-121°C
Appearance (Color)Pale cream to cream to pale brown or pale yellow to orange
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only