(R)-(-)-alpha-Methoxyphenylacetic acid, 99%, Thermo Scientific Chemicals
(R)-(-)-alpha-Methoxyphenylacetic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(R)-(-)-alpha-Methoxyphenylacetic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number L08719.03
also known as L08719-03
Price (USD)/ Each
104.65
Online exclusive
116.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
104.65
Online exclusive
116.00 
Save 11.35 (10%)
Add to cart
(R)-(-)-alpha-Methoxyphenylacetic acid, 99%, Thermo Scientific Chemicals
Catalog numberL08719.03
Price (USD)/ Each
104.65
Online exclusive
116.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS3966-32-3
IUPAC Name(2R)-2-methoxy-2-phenylacetate
Molecular FormulaC9H9O3
InChI KeyDIWVBIXQCNRCFE-MRVPVSSYSA-M
SMILESCO[C@@H](C([O-])=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5% (non-U.S. specification)
Assay (Silylated GC)≥98.5% (non-U.S. specification)
Assay from Suppliers CofA≥98.5% (U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only