It is employed as a intermediate for organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as a intermediate for organic synthesis.
Solubility
Insoluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
W F Lau; and N P Das. In vitro modulation of rat adipocyte ghost membrane fluidity by cholesterol oxysterols. Experientia. Basel. 1995, 5 (7), 731-737.
P P Nair; N Turjman; G T Goodman; C Guidry; B M Calkins. Diet, nutrition intake, and metabolism in populations at high and low risk for colon cancer. Metabolism of neutral sterols. American Journal of Clinical Nutrition. 1984, 40(4 Suppl)931-936.