Thermo Scientific Chemicals

(S)-(+)-alpha-Methoxyphenylacetic acid, 99%, Thermo Scientific Chemicals

Catalog number: L08748.MD
250 mg, Each
Thermo Scientific Chemicals

(S)-(+)-alpha-Methoxyphenylacetic acid, 99%, Thermo Scientific Chemicals

Catalog number: L08748.MD
250 mg, Each
Quantity
Catalog number: L08748.MD
also known as L08748-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
26164-26-1
IUPAC Name
(2S)-2-methoxy-2-phenylacetate
Molecular Formula
C9H9O3
InChI Key
DIWVBIXQCNRCFE-QMMMGPOBSA-M
SMILES
CO[C@H](C([O-])=O)C1=CC=CC=C1
Comment
Material sourced in the U.S. and in other countries
Optical Rotation
146.5° ± 3.0° (c=0.5 in ethanol, non-U.S. sourced material)
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5% (non-U.S. sourced material)
Assay from Supplier's CofA
≥98.5% (U.S. sourced material)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text