Thermo Scientific Chemicals

3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride, 98%, Thermo Scientific Chemicals

Catalog number: L08750.22
100 g, Each
Thermo Scientific Chemicals

3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride, 98%, Thermo Scientific Chemicals

Catalog number: L08750.22
100 g, Each
Quantity
Catalog number: L08750.22
also known as L08750-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4568-71-2
IUPAC Name
3-benzyl-5-(2-hydroxyethyl)-4-methyl-1,3-thiazol-3-ium chloride
Molecular Formula
C13H16ClNOS
InChI Key
IWSVLBKHBJGMAA-UHFFFAOYSA-M
SMILES
[Cl-].CC1=C(CCO)SC=[N+]1CC1=CC=CC=C1
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
≤0.5%

Description

3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride, aliphatic aldehydes undergo the acyloin condensation in the presence of this catalyst and mild base. It acts as a catalyst for the addition of aliphatic and heterocyclic aldehydes to α,β-unsaturated ketones, nitriles and esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride, aliphatic aldehydes undergo the acyloin condensation in the presence of this catalyst and mild base. It acts as a catalyst for the addition of aliphatic and heterocyclic aldehydes to α,β-unsaturated ketones, nitriles and esters.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from water and oxidizing agents. Protect from humidity and water. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text