Thermo Scientific Chemicals

Phthaloyl chloride, 94%, Thermo Scientific Chemicals

Catalog number: L08753.36
500 g, Each
Thermo Scientific Chemicals

Phthaloyl chloride, 94%, Thermo Scientific Chemicals

Catalog number: L08753.36
500 g, Each
Quantity
Catalog number: L08753.36
also known as L08753-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
88-95-9
IUPAC Name
benzene-1,2-dicarbonyl dichloride
Molecular Formula
C8H4Cl2O2
InChI Key
FYXKZNLBZKRYSS-UHFFFAOYSA-N
SMILES
ClC(=O)C1=CC=CC=C1C(Cl)=O
Form
Liquid
Assay from Supplier's CofA
≥93.0%
Appearance (Color)
Clear colorless to yellow

Description

Phthaloyl chloride is used for the preparation of volatile acid chlorides from the acids. It acts as a trapping agent for the nitrite ion in the fluorodenitration of aromatic substrates with potassium fluoride. It serves as an additive in polymers and fibers to improve the color stability. It is used as an ingredient in ceramic mold binders, as a stabilizer for polymers and in reverse osmosis membranes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phthaloyl chloride is used for the preparation of volatile acid chlorides from the acids. It acts as a trapping agent for the nitrite ion in the fluorodenitration of aromatic substrates with potassium fluoride. It serves as an additive in polymers and fibers to improve the color stability. It is used as an ingredient in ceramic mold binders, as a stabilizer for polymers and in reverse osmosis membranes.

Solubility
Miscible with ether.

Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with water, alcohols oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text