Thermo Scientific Chemicals

1-Eicosanol, 96%, Thermo Scientific Chemicals

Catalog number: L08807.14
25 g, Each
Thermo Scientific Chemicals

1-Eicosanol, 96%, Thermo Scientific Chemicals

Catalog number: L08807.14
25 g, Each
Quantity
Catalog number: L08807.14
also known as L08807-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
629-96-9
IUPAC Name
icosan-1-ol
Molecular Formula
C20H42O
InChI Key
BTFJIXJJCSYFAL-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCCCCCO
Appearance (Color)
White
Form
Powder or lumps or flakes or pellets
Assay (GC)
≥95.0%
Melting Point (clear melt)
60.0-70.0?C

Description

1-Eicosanol has been used in gradient HPLC-charged aerosol detection method for the detection of different lipids, as internal standard during determination of policosanol?s high molecular weight alcohols (policosanol?s fatty alcohols) in fish oil by GC. Also used as a non-ionic surfactant in cosmetics, used in lubricants, rubber, plastics, textiles and as an emulsion stabilizer and viscosity increasing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Eicosanol has been used in gradient HPLC-charged aerosol detection method for the detection of different lipids, as internal standard during determination of policosanol′s high molecular weight alcohols (policosanol′s fatty alcohols) in fish oil by GC. Also used as a non-ionic surfactant in cosmetics, used in lubricants, rubber, plastics, textiles and as an emulsion stabilizer and viscosity increasing agent.

Solubility
Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text