Thermo Scientific Chemicals

N(alpha)-Boc-D-histidine, 98+%

Catalog number: L08810.03
1 g, Each
Thermo Scientific Chemicals

N(alpha)-Boc-D-histidine, 98+%

Catalog number: L08810.03
1 g, Each
Quantity
Catalog number: L08810.03
also known as L08810-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
50654-94-9
IUPAC Name
(2R)-2-{[(tert-butoxy)carbonyl]amino}-3-(1H-imidazol-5-yl)propanoate
Molecular Formula
C11H16N3O4
InChI Key
AYMLQYFMYHISQO-MRVPVSSYSA-M
SMILES
CC(C)(C)OC(=O)N[C@H](CC1=CN=CN1)C([O-])=O
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder
Optical Rotation
-26.0? ? 1.0? (c=1 in methanol)
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text