Dibenzyl phosphate, 98%, Thermo Scientific Chemicals
Dibenzyl phosphate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dibenzyl phosphate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number L08813.14
also known as L08813-14
Price (USD)/ Each
279.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
279.00
Add to cart
Dibenzyl phosphate, 98%, Thermo Scientific Chemicals
Catalog numberL08813.14
Price (USD)/ Each
279.00
-
Add to cart
Chemical Identifiers
CAS1623-08-1
IUPAC Namebis(benzyloxy)phosphinic acid
Molecular FormulaC14H15O4P
InChI KeyHDFFVHSMHLDSLO-UHFFFAOYSA-N
SMILESOP(=O)(OCC1=CC=CC=C1)OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder or lumps or fused solid
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
Melting Point (clear melt)76.0-83.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only