Thermo Scientific Chemicals

2,2-Di-n-propylacetyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: L08852.06
5 g, Each
Thermo Scientific Chemicals

2,2-Di-n-propylacetyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: L08852.06
5 g, Each
Quantity
Catalog number: L08852.06
also known as L08852-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2936-08-5
IUPAC Name
2-propylpentanoyl chloride
Molecular Formula
C8H15ClO
InChI Key
PITHYUDHKJKJNQ-UHFFFAOYSA-N
SMILES
CCCC(CCC)C(Cl)=O
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.4300-1.4340 @ 20°C

Description

2,2-Di-n-propylacetyl chloride is used to produce 1,2-bis-(2-propyl-valerylamino)-ethan.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Di-n-propylacetyl chloride is used to produce 1,2-bis-(2-propyl-valerylamino)-ethan.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Incompatible materials are Oxidizing agents and bases. Store away from moisture and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text