Thermo Scientific Chemicals

N,N-Diisopropylformamide, 98%, Thermo Scientific Chemicals

Catalog number: L08857.06
5 g, Each
Thermo Scientific Chemicals

N,N-Diisopropylformamide, 98%, Thermo Scientific Chemicals

Catalog number: L08857.06
5 g, Each
Quantity
Catalog number: L08857.06
also known as L08857-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2700-30-3
IUPAC Name
N,N-bis(propan-2-yl)formamide
Molecular Formula
C7H15NO
InChI Key
UNBDDZDKBWPHAX-UHFFFAOYSA-N
SMILES
CC(C)N(C=O)C(C)C
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.4340-1.4380 @ 20?C

Description

It is employed as a intermediate for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for organic synthesis.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text