3-Ethylbenzothiazolium bromide, 98%, Thermo Scientific Chemicals
3-Ethylbenzothiazolium bromide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Ethylbenzothiazolium bromide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number L08903.18
also known as L08903-18
Price (USD)/ Each
238.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
238.00
Add to cart
3-Ethylbenzothiazolium bromide, 98%, Thermo Scientific Chemicals
Catalog numberL08903.18
Price (USD)/ Each
238.00
-
Add to cart
Chemical Identifiers
CAS32446-47-2
IUPAC Name3-ethyl-1,3-benzothiazol-3-ium bromide
Molecular FormulaC9H10BrNS
InChI KeyXBWYYTURPBHPFJ-UHFFFAOYSA-M
SMILES[Br-].CC[N+]1=CSC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (Titration ex Bromide)≥97.5 to ≤102.5%
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only