N-Benzyloxycarbonyl-6-aminohexanoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-Benzyloxycarbonyl-6-aminohexanoic acid, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L08997.06
also known as L08997-06
Price (USD)/ Each
54.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
54.00
Add to cart
N-Benzyloxycarbonyl-6-aminohexanoic acid, 98%, Thermo Scientific Chemicals
Catalog numberL08997.06
Price (USD)/ Each
54.00
-
Add to cart
Chemical Identifiers
CAS1947-00-8
IUPAC Name6-{[(benzyloxy)carbonyl]amino}hexanoic acid
Molecular FormulaC14H19NO4
InChI KeyRXQDBVWDABAAHL-UHFFFAOYSA-N
SMILESOC(=O)CCCCCNC(=O)OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (HPLC)≥97.5%
FormPowder
Melting Point53-58?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only