Thermo Scientific Chemicals

N-Benzyloxycarbonyl-6-aminohexanoic acid, 98%, Thermo Scientific Chemicals

Catalog number: L08997.14
25 g, Each
Thermo Scientific Chemicals

N-Benzyloxycarbonyl-6-aminohexanoic acid, 98%, Thermo Scientific Chemicals

Catalog number: L08997.14
25 g, Each
Quantity
Catalog number: L08997.14
also known as L08997-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1947-00-8
IUPAC Name
6-{[(benzyloxy)carbonyl]amino}hexanoic acid
Molecular Formula
C14H19NO4
InChI Key
RXQDBVWDABAAHL-UHFFFAOYSA-N
SMILES
OC(=O)CCCCCNC(=O)OCC1=CC=CC=C1
Appearance (Color)
White to pale cream
Assay (HPLC)
≥97.5%
Form
Powder
Melting Point
53-58?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text