Thermo Scientific Chemicals

Aminomalononitrile p-toluenesulfonate, 97+%, Thermo Scientific Chemicals

Catalog number: L09001.14
25 g, Each
Thermo Scientific Chemicals

Aminomalononitrile p-toluenesulfonate, 97+%, Thermo Scientific Chemicals

Catalog number: L09001.14
25 g, Each
Quantity
Catalog number: L09001.14
also known as L09001-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5098-14-6
IUPAC Name
2-aminopropanedinitrile; 4-methylbenzene-1-sulfonic acid
Molecular Formula
C10H11N3O3S
InChI Key
MEUWQVWJLLBVQI-UHFFFAOYSA-N
SMILES
NC(C#N)C#N.CC1=CC=C(C=C1)S(O)(=O)=O
Loss on Drying
<1.0% (105?C/ constant weight)
Form
powder
Appearance (Color)
White to cream
Assay (Aqueous acid-base Titration)
> 97.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text