4-Dimethylaminophenyl isothiocyanate, 97%, Thermo Scientific Chemicals
4-Dimethylaminophenyl isothiocyanate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Dimethylaminophenyl isothiocyanate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
Catalog number L09011.09
also known as L09011-09
Price (USD)/ Each
207.65
Online exclusive
230.00 
Save 22.35 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
207.65
Online exclusive
230.00 
Save 22.35 (10%)
Add to cart
4-Dimethylaminophenyl isothiocyanate, 97%, Thermo Scientific Chemicals
Catalog numberL09011.09
Price (USD)/ Each
207.65
Online exclusive
230.00 
Save 22.35 (10%)
-
Add to cart
Chemical Identifiers
CAS2131-64-8
IUPAC Name4-isothiocyanato-N,N-dimethylaniline
Molecular FormulaC9H10N2S
InChI KeyHRDJPEMAGYHSJR-UHFFFAOYSA-N
SMILESCN(C)C1=CC=C(C=C1)N=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to yellow to green
Assay (GC)≥96.0%
Melting Point (clear melt)63.0-73.0?C
FormCrystals or powder or crystalline powder or lumps or fused solid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only