Thermo Scientific Chemicals

Ethyl isobutyrylacetate, 94%, Thermo Scientific Chemicals

Catalog number: L09016.14
25 g, Each
Thermo Scientific Chemicals

Ethyl isobutyrylacetate, 94%, Thermo Scientific Chemicals

Catalog number: L09016.14
25 g, Each
Quantity
Catalog number: L09016.14
also known as L09016-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7152-15-0
IUPAC Name
ethyl 4-methyl-3-oxopentanoate
Molecular Formula
C8H14O3
InChI Key
XCLDSQRVMMXWMS-UHFFFAOYSA-N
SMILES
CCOC(=O)CC(=O)C(C)C
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.4225-1.4300 @ 20?C
Form
Liquid
Assay (Silylated GC)
>92.5%

Description

Ethyl Isobutyrylacetate is used in the synthesis of piperazine derivatives as possible multireceptor atypical antipsychotics, affecting dopamine and serotonin receptor properties . Also used in the synthesis of pyrazinecarboxamide DGAT1 (diacylglycerol acyltransferase 1) inhibitors used in the treatment of obesity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl Isobutyrylacetate is used in the synthesis of piperazine derivatives as possible multireceptor atypical antipsychotics, affecting dopamine and serotonin receptor properties . Also used in the synthesis of pyrazinecarboxamide DGAT1 (diacylglycerol acyltransferase 1) inhibitors used in the treatment of obesity.

Solubility
Soluble in chloroform and methanol.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text