Thermo Scientific Chemicals

DL-Lactide, 99%, Thermo Scientific Chemicals

Catalog number: L09026.18
50 g, Each
Thermo Scientific Chemicals

DL-Lactide, 99%, Thermo Scientific Chemicals

Catalog number: L09026.18
50 g, Each
Quantity
Catalog number: L09026.18
also known as L09026-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
95-96-5
IUPAC Name
3,6-dimethyl-1,4-dioxane-2,5-dione
Molecular Formula
C6H8O4
InChI Key
JJTUDXZGHPGLLC-UHFFFAOYNA-N
SMILES
CC1OC(=O)C(C)OC1=O
Appearance (Color)
White
Assay (GC)
≥98.5%
Form
Crystals or crystalline powder
Melting Point (clear melt)
119-128?C
Identification (FTIR)
Conforms

Description

DL-Lactide is used to produce 2-hydroxy-propionic acid 1-(1-phenyl-ethoxycarbonyl)-ethyl ester. It acts as an important raw material and an intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyes. It is involved in enzymatic alcoholysis to produce both alkyl (R)-lactates and alkyl (S,S)-lactyllactates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Lactide is used to produce 2-hydroxy-propionic acid 1-(1-phenyl-ethoxycarbonyl)-ethyl ester. It acts as an important raw material and an intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyes. It is involved in enzymatic alcoholysis to produce both alkyl (R)-lactates and alkyl (S,S)-lactyllactates.

Solubility
Soluble in chloroform and methanol. Slightly soluble in benzene.

Notes
Moisture senstive. Store in a cool place. Incompatible with acids, bases and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text