Thermo Scientific Chemicals

L-Lactide, 98+%, Thermo Scientific Chemicals

Catalog number: L09031.14
25 g, Each
Thermo Scientific Chemicals

L-Lactide, 98+%, Thermo Scientific Chemicals

Catalog number: L09031.14
25 g, Each
Quantity
Catalog number: L09031.14
also known as L09031-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4511-42-6
IUPAC Name
3,6-dimethyl-1,4-dioxane-2,5-dione
Molecular Formula
C6H8O4
InChI Key
JJTUDXZGHPGLLC-UHFFFAOYNA-N
SMILES
CC1OC(=O)C(C)OC1=O
Identification (FTIR)
Conforms
Optical Rotation
-270? ? 6? (c = 1 in chloroform)
Form
Crystalline powder
Melting Point (clear melt)
95-100?C
Appearance (Color)
White

Description

L-Lactide upon polymerization gives poly(lactic acid), which finds many useful properties like biodegradable and nontoxic to the human body and the environment. Its polymer, poly-l-lactide is utilized in the repairing of orbital posterior floor fracture. Further, it is used as biomedical materials for tissue regeneration, matrices for drug delivery systems and alternatives for commercial polymeric materials to reduce the impact on the environment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in chloroform, and methanol. Slightly soluble in benzene.

Notes
Moisture sensitive. Store in a cool place. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text