Thermo Scientific Chemicals

DL-Homocysteine thiolactone hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: L09077.14
25 g, Each
Thermo Scientific Chemicals

DL-Homocysteine thiolactone hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: L09077.14
25 g, Each
Quantity
Catalog number: L09077.14
also known as L09077-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6038-19-3
IUPAC Name
hydrogen 3-aminothiolan-2-one chloride
Molecular Formula
C4H8ClNOS
InChI Key
ZSEGSUBKDDEALH-UHFFFAOYNA-N
SMILES
[H+].[Cl-].NC1CCSC1=O
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%
Appearance (Color)
White to pale yellow or cream

Description

DL-Homocysteine thiolactone hydrochloride is used as an intermediate of erdosteine and citiolone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Homocysteine thiolactone hydrochloride is used as an intermediate of erdosteine and citiolone.

Solubility
Soluble in water (740g/L). Soluble in dimethyl sulfoxide and methanol.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text