Thermo Scientific Chemicals

N(alpha)-Benzyloxycarbonyl-L-histidine, 98+%, Thermo Scientific Chemicals

Catalog number: L09114.06
5 g, Each
Thermo Scientific Chemicals

N(alpha)-Benzyloxycarbonyl-L-histidine, 98+%, Thermo Scientific Chemicals

Catalog number: L09114.06
5 g, Each
Quantity
Catalog number: L09114.06
also known as L09114-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14997-58-1
IUPAC Name
2-{[(benzyloxy)carbonyl]amino}-3-(1H-imidazol-5-yl)propanoic acid
Molecular Formula
C14H15N3O4
InChI Key
WCOJOHPAKJFUDF-UHFFFAOYNA-N
SMILES
OC(=O)C(CC1=CN=CN1)NC(=O)OCC1=CC=CC=C1
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Form
Crystals or powder or crystalline powder
Optical Rotation
-24.2 ± 0.1? (C=6 in 6M HCl)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text