3,3'-Diaminobenzophenone, 90%, Thermo Scientific Chemicals
3,3'-Diaminobenzophenone, 90%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,3'-Diaminobenzophenone, 90%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L09128.03
also known as L09128-03
Price (USD)/ Each
69.10
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
69.10
Add to cart
3,3'-Diaminobenzophenone, 90%, Thermo Scientific Chemicals
Catalog numberL09128.03
Price (USD)/ Each
69.10
-
Add to cart
Chemical Identifiers
CAS611-79-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to pale brown
FormPowder
Assay (GC)≥88.0%
Assay (Non-aqueous acid-base Titration)≥88.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only