Thermo Scientific Chemicals

6-Methyl-3,5-heptadien-2-one, 97%, Thermo Scientific Chemicals

Catalog number: L09169.14
25 g, Each
Thermo Scientific Chemicals

6-Methyl-3,5-heptadien-2-one, 97%, Thermo Scientific Chemicals

Catalog number: L09169.14
25 g, Each
Quantity
Catalog number: L09169.14
also known as L09169-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1604-28-0
IUPAC Name
(3E)-6-methylhepta-3,5-dien-2-one
Molecular Formula
C8H12O
InChI Key
KSKXSFZGARKWOW-GQCTYLIASA-N
SMILES
CC(C)=CC=CC(C)=O
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.5315-1.5365 @ 20?C

Description

6-Methyl-3,5-heptadien-2-one is used in the preparation of (RS)-(E)-2,6-dimethyl-1,2-epoxy-3,5-heptadiene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methyl-3,5-heptadien-2-one is used in the preparation of (RS)-(E)-2,6-dimethyl-1,2-epoxy-3,5-heptadiene.

Solubility
Miscible with alcohol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text