Thermo Scientific Chemicals

3-Phenylpropiophenone, 98%, Thermo Scientific Chemicals

Catalog number: L09173.18
50 g, Each
Thermo Scientific Chemicals

3-Phenylpropiophenone, 98%, Thermo Scientific Chemicals

Catalog number: L09173.18
50 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L09173.18
also known as L09173-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1083-30-3
IUPAC Name
1,3-diphenylpropan-1-one
Molecular Formula
C15H14O
InChI Key
QGGZBXOADPVUPN-UHFFFAOYSA-N
SMILES
O=C(CCC1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White to pale brown
Form
Crystalline powder
Assay (GC)
>97.5%
Clarity
(5% w/v solution in acetone) : Transparent to almost transparent by JIS K 8001 5.2

Description

3-Phenylpropiophenone is used to produce 2-bromo-1,3-diphenyl-propan-1-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Phenylpropiophenone is used to produce 2-bromo-1,3-diphenyl-propan-1-one.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text