4-n-Pentylbenzaldehyde, 95%, Thermo Scientific Chemicals
4-n-Pentylbenzaldehyde, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-n-Pentylbenzaldehyde, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L09208.03
also known as L09208-03
Price (USD)/ Each
55.40
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
55.40
Add to cart
4-n-Pentylbenzaldehyde, 95%, Thermo Scientific Chemicals
Catalog numberL09208.03
Price (USD)/ Each
55.40
-
Add to cart
Chemical Identifiers
CAS6853-57-2
IUPAC Name4-pentylbenzaldehyde
Molecular FormulaC12H16O
InChI KeyNQVZPRUSNWNSQH-UHFFFAOYSA-N
SMILESCCCCCC1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to white to pale yellow
Free acid (titration)≤3.75%
FormLiquid
Assay (GC)≥94.0%
Refractive Index1.5185-1.5245 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only