Thermo Scientific Chemicals

1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol, 98%, Thermo Scientific Chemicals

Catalog number: L09245.06
5 g, Each
Thermo Scientific Chemicals

1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol, 98%, Thermo Scientific Chemicals

Catalog number: L09245.06
5 g, Each
Quantity
Catalog number: L09245.06
also known as L09245-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1515-14-6
IUPAC Name
1,1,1,3,3,3-hexafluoro-2-methylpropan-2-ol
Molecular Formula
C4H4F6O
InChI Key
FQDXJYBXPOMIBX-UHFFFAOYSA-N
SMILES
CC(O)(C(F)(F)F)C(F)(F)F
Appearance (Color)
Clear colorless to pale yellow or pale brown
Identification (FTIR)
Conforms
Form
Liquid
Comment
Material sourced in UK and US
Refractive Index
<1.3000 @ 20?C (UK Sourced Material)

Description

1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol is used as a solvent. It is used in the preparation of volatile sodium yttrium and zirconium fluoroalkoxides. It is also used in the synthesis of precursors to per- and polyfluoroethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol is used as a solvent. It is used in the preparation of volatile sodium yttrium and zirconium fluoroalkoxides. It is also used in the synthesis of precursors to per- and polyfluoroethers.

Solubility
Slightly soluble in water.

Notes
Incompatible materials are strong acids, strong bases, strong oxidizing agents, strong reducing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text