Thermo Scientific Chemicals

N,N,3,5-Tetramethylaniline, 98%, Thermo Scientific Chemicals

Catalog number: L09266.14
25 g, Each
Thermo Scientific Chemicals

N,N,3,5-Tetramethylaniline, 98%, Thermo Scientific Chemicals

Catalog number: L09266.14
25 g, Each
Quantity
Catalog number: L09266.14
also known as L09266-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4913-13-7
IUPAC Name
N,N,3,5-tetramethylaniline
Molecular Formula
C10H15N
InChI Key
NBFRQCOZERNGEX-UHFFFAOYSA-N
SMILES
CN(C)C1=CC(C)=CC(C)=C1
Form
or pale brown liquid
Refractive Index
1.5420-1.5460
Assay (GC)
>97.5%
Appearance (Color)
Clear, colourless to yellow

Description

N,N,3,5-Tetramethylaniline was used in rapid quenching of 1,8-dihydroxyanthraquinone (DHAQ). It was also used as a catalyst in the polymeric synthesis of glycol methacrylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N,3,5-Tetramethylaniline was used in rapid quenching of 1,8-dihydroxyanthraquinone (DHAQ). It was also used as a catalyst in the polymeric synthesis of glycol methacrylate.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text